Learn More
Biomatik Corporation Human Inhibin B (INHB) ELISA Kit
Supplier:  Biomatik Corporation EKU10140
Encompass_Preferred
Human Inhibin B (INHB) ELISA Kit for detecting INHB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05111; P09529. Gene ID: 3623; 3625. Target Synonyms: INH-B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6070