Learn More
Biomatik Corporation Human Inhibin Beta A (INHbA) ELISA Kit
Supplier:  Biomatik Corporation EKU08710
Encompass_Preferred
Human Inhibin Beta A (INHbA) ELISA Kit for detecting INHbA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.17-500pg/ml. Sensitivity: 2.39pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P08476. Gene ID: 3624. Target Synonyms: INH-BA; INHB-A; EDF; FRP; Activin A,Activin AB Alpha Polypeptide; Activin beta-A chain; Erythroid differentiation protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4855