Learn More
Sino Biological Recombinant Human Inhibin beta B Protein (His Tag)
Supplier:  Sino Biological 10814H08H50
Encompass_Preferred
A DNA sequence encoding the pro form of human INHBB (NP_002184.2) (Met 1-Ala 407) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6867