Learn More
Biomatik Corporation Human Inhibitory Subunit Of NF Kappa B Alpha (IkBa) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10935
Encompass_Preferred
Human Inhibitory Subunit Of NF Kappa B Alpha (IkBa) High-sensitive ELISA Kit for detecting IkBa in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P25963. Gene ID: 4792. Target Synonyms: NFKBIA; IKB-A; MAD-3; NFKBI; Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells Inhibitor,Alpha; Major histocompatibility complex enhancer-binding protein MAD3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8265