Learn More
Sino Biological Pro-Insulin C-peptide Antibody, Mouse MAb
Supplier:  Sino Biological 110121MM15100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with a synthetic peptide corresponding to the Human Pro-Insulin C-peptide. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8842