Learn More
Sino Biological Recombinant Anti-Insulin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11038CR007100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD28 (rh Insulin; Catalog# 11038-HNAY; P01308; Phe25-Lys53 & Gly90-Asn110).
Catalog No. 50-240-9857