Learn More
Sino Biological Anti-Insulin Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11038MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Insulin (rh Insulin; Catalog#11038-HNAY; P01308; Phe25-Lys53 & Gly90-Asn110). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-7909