Learn More
Sino Biological Human INSR MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11086MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Insulin Receptor/INSR/CD220 (rh Insulin Receptor/INSR/CD220; Catalog#11086-H08H; NP_001073285.1; Met1-Lys944). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1554