Learn More
Sino Biological Anti-Insulin Receptor Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 11086MM01P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Insulin Receptor/INSR/CD220 (rh Insulin Receptor/INSR/CD220; Catalog#11086-H08H; NP_001073285.1; Met1-Lys944) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-4842