Learn More
Sino Biological Anti-Insulin Receptor Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 11081MM01H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human INSR / Insulin Receptor extracellular domain (rh INSR; Catalog#11081-H08H; NP_000199.2; Met 1-Lys 956). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-255-4867