Learn More
Sino Biological Human INSR PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50010R485A100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Insulin Receptor / CD220 / INSR (rh Insulin Receptor / CD220 / INSR; Catalog#11081-H08H; NP_000199.2; Met 1-Lys 956). Insulin Receptor / CD220 / INSR specific IgG was purified by Human Insulin Receptor / CD220 / INSR affinity chromatography.
Catalog No. 50-237-4486