Learn More
Sino Biological Human INSR PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11081RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human INSR / Insulin Receptor extracellular domain (rh INSR; Catalog#11081-H08H; NP_000199.2; Met 1-Lys 956). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-1438