Learn More
Sino Biological Recombinant Human Insulin Receptor Protein (ECD, long isoform, His Tag), HPLC-verified
Supplier:  Sino Biological 11081H08H1
Encompass_Preferred
A DNA sequence encoding the human INSR isoform long (NP_000199.2) extracellular domain (Met1-Lys956) was expressed fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4871