Learn More
Sino Biological Recombinant Human Insulin Receptor Protein (long isoform, His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 11081H08HB20
Encompass_Preferred
A DNA sequence encoding the human INSR isoform long (NP_000199.2) extracellular domain (Met1-Lys956) was expressed, fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4870