Learn More
Sino Biological Human Insulin Receptor Protein (short isoform, His & AVI Tag), Biotinylated
Supplier:  Sino Biological 11086H49HB20
Encompass_Preferred
A DNA sequence encoding the Human INSR (NP_001073285.1) (Met1-Lys944) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4843