Learn More
Raybiotech Inc Human Insulin ELISA Kit
Supplier:  Raybiotech Inc ELHINSULIN1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 4 uIU/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human BDNF, BLC, ENA-78, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, I-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TNF-alpha, TNF-beta, TPO, VEGF.
Catalog No. 50-194-9465