Learn More
Mercodia Inc Insulin ELISA, Human, High Specificity, Serum Plasma, 96 Wells
SDP
Supplier:  Mercodia Inc 10111301
Encompass_Preferred
A high quality enzyme immunoassay for specific quantification of human insulin in serum, plasma, or cell culture media. The Mercodia insulin assays have little or no cross-reactivity to insulin analogs, rat or mouse insulin, which enables distinguishing endogenous human insulin, both in patients undergoing treatment with insulin analogs, and in rat or mouse models.
Catalog No. NC9597838