Learn More
Alpco Ltd Insulin ELISA
SDP
Supplier:  Alpco Ltd 80INSHUE01.1
Encompass_Preferred
The Insulin ELISA quantifies the concentration of insulin protein in human serum or plasma samples. FDA Class I Device. For In Vitro Diagnostic Use.
  • Only 25 µL of sample is sufficient for the assay
  • Superior sensitivity of 0.399 µIU/mL
  • Dynamic range of 3.0-200 µIU/mL
Catalog No. NC0038324