Learn More
Biomatik Corporation Human Insulin (INS) CLIA Kit
Supplier:  Biomatik Corporation EKU08619
Encompass_Preferred
Human Insulin (INS) CLIA Kit for detecting INS in serum, plasma, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 39.1-10,000pg/ml. Sensitivity: 15.1pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P01308. Gene ID: 3630. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6084