Learn More
Biomatik Corporation Human Insulin Like Growth Factor Binding Protein 7 (IGFBP7) CLIA Kit
Supplier:  Biomatik Corporation EKU09924
Encompass_Preferred
Human Insulin Like Growth Factor Binding Protein 7 (IGFBP7) CLIA Kit for detecting IGFBP7 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.82-600ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q16270. Gene ID: 3490. Target Synonyms: FSTL2; IGFBP-7v; MAC25; PSF; TAF; IGFBP-rP1; MAC25 protein; Prostacyclin-stimulating factor; PGI2-stimulating factor; Tumor-derived adhesion factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4388