Learn More
Biomatik Corporation Human Insulin Like Growth Factor Binding Protein 4 (IGFBP4) CLIA Kit
Supplier:  Biomatik Corporation EKU08789
Encompass_Preferred
Human Insulin Like Growth Factor Binding Protein 4 (IGFBP4) CLIA Kit for detecting IGFBP4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.55-400ng/ml. Sensitivity: 0.23ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P22692. Gene ID: 3487. Target Synonyms: BP4; HT29-IGFBP; IBP4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6725