Learn More
Biomatik Corporation Human Insulin Like Protein 5 (INSL5) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10556
Encompass_Preferred
Human Insulin Like Protein 5 (INSL5) Wide-range ELISA Kit for detecting INSL5 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y5Q6. Gene ID: 10022. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4979