Learn More
Biomatik Corporation Human Insulin Receptor Substrate 2 (IRS2) ELISA Kit
Supplier:  Biomatik Corporation EKU09179
Encompass_Preferred
Human Insulin Receptor Substrate 2 (IRS2) ELISA Kit for detecting IRS2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y4H2. Gene ID: 8660. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4766