Learn More
IBL America Angiotensinogen (Intact)(Human) ELISA
Supplier:  IBL America 27742
Encompass_Preferred
Solid phase sandwich ELISA for the determination of human Intact Angiotensinogen ELISA in serum, EDTA-plasma, and urine. For research use only, not for use in diagnostic procedures.
Catalog No. 50-212-0356