Learn More
ACROBiosystems Integrin α 4 β 7 : MAdCAM-1 [Biotinylated] Inhibitor Screening ELISA Kit
Catalog No. 502329128
Encompass_Preferred
The integrin α4β7 is heterodimeric cell surface receptors expressed on most leukocytes. Mucosal addressing cell adhesion molecule 1(MAdCAM-1) is an exclusive ligand for α4β7 integrin. The interaction of integrin α4β7 with addressin mucosal addressin cell adhesion molecule-1 (MAdCAM-1) is central to IBD pathogenesis.
Catalog No. 50-232-9128 Supplier ACROBiosystems Supplier No. EP15596TESTS
The integrin α4β7 is heterodimeric cell surface receptors expressed on most leukocytes. Mucosal addressing cell adhesion molecule 1(MAdCAM-1) is an exclusive ligand for α4β7 integrin. The interaction of integrin α4β7 with addressin mucosal addressin cell adhesion molecule-1 (MAdCAM-1) is central to IBD pathogenesis.