Learn More
ACROBiosystems Human Integrin alpha 1 100ug
Catalog No. 502534565
Encompass_Preferred
Human Integrin alpha 1 Protein His Tag (IN1-H52H3) is expressed from human 293 cells (HEK293). It contains AA Phe 29-Pro1141 (Accession # P56199-1).
Catalog No. 50-253-4565 Supplier ACROBiosystems Supplier No. IN1H52H3100UG
Human Integrin alpha 1 Protein His Tag (IN1-H52H3) is expressed from human 293 cells (HEK293). It contains AA Phe 29-Pro1141 (Accession # P56199-1).