Learn More
Biomatik Corporation Human Integrin Alpha 10 (ITGa10) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU11599
Encompass_Preferred
Human Integrin Alpha 10 (ITGa10) High-sensitive ELISA Kit for detecting ITGa10 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75578. Gene ID: 8515. Target Synonyms: ITG-A10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8122