Learn More
ACROBiosystems ACROBiosystems Human Integrin alpha 6 beta 1 (ITGA6&ITGB1) Heterodimer Protein, His Tag&Tag Free
Supplier:  ACROBiosystems IT1H52W71MG
Encompass_Preferred
Recombinant Protein;1MG;Human ITGA6&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-H52W7) is expressed from human 293 cells (HEK293). It contains AA Phe 24 - Gly 1012 (ITGA6) & Gln 21 - Asp 728 (ITGB1) (Accession # P23229-2 (ITGA6) & P05556-1 (ITGB1)).;Human ITGA6&ITGB1 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGA6 and ITGB1, has a calculated MW of 116.8 kDa (ITGA6) and 83.7 kDa (ITGB1). Subunit ITGA6 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The non-reducing (NR) protein migrates as 140-150 kDa (ITGA6) and 100-118 kDa (ITGB1) respectively due to glycosylation.
Catalog No. 50-210-6686