Learn More
Biomatik Corporation Human Integrin Beta 1 (ITGb1) Instant ELISA Kit
Supplier:  Biomatik Corporation EKU11002
Encompass_Preferred
Human Integrin Beta 1 (ITGb1) Instant ELISA Kit for detecting ITGb1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 1h, 10min. Accession Number: P05556. Gene ID: 3688. Target Synonyms: CD29; MSK12; ITG-B1; FNRB; GPIIA; MDF2; MSK12; VLAB; Fibronectin Receptor Beta Polypeptide; Antigen CD29 Includes MDF2; Glycoprotein IIa; VLA-4 subunit beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6073