Learn More
Biomatik Corporation Human Integrin Beta 6 (ITGb6) CLIA Kit
Supplier:  Biomatik Corporation EKU09780
Encompass_Preferred
Human Integrin Beta 6 (ITGb6) CLIA Kit for detecting ITGb6 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 4.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P18564. Gene ID: 3694. Target Synonyms: ITG-B6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8436