Learn More
Bioworld HUMAN INTERCELLULAR ADHESIO
Supplier:  Bioworld 220603621
Encompass_Preferred
HUMAN INTERCELLULAR ADHESION MOLECULE-1 (RECOMBINANT): Recombinant Human Intercellular Adhesion Molecule-1 (ICAM-1) Source: E.Coli Background : ICAM-1 (CD54) is a single chain membrane glycoprotein expressed on the surface of a varie...
Catalog No. 50-253-459