Learn More
American Research Products Inc Human intercellular adhesion m
Catalog No. 5000328796
Encompass
Human intercellular adhesion molecule 3 ICAM-3 ELISA Kit from CUSABIO Detection Range 31 25 pg/ml - 2000 pg/ml Sensitivity 7 8 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28796 Supplier American Research Products Inc Supplier No. CSBE07949H
Human intercellular adhesion molecule 3 ICAM-3 ELISA Kit from CUSABIO Detection Range 31 25 pg/ml - 2000 pg/ml Sensitivity 7 8 pg/ml Sample serum plasma tissue homogenates Method Sandwich