Learn More
Biomatik Corporation Human Intercellular Adhesion Molecule 4 (ICAM4) ELISA Kit
Supplier:  Biomatik Corporation EKU05066
Encompass_Preferred
Human Intercellular Adhesion Molecule 4 (ICAM4) ELISA Kit for detecting ICAM4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14773. Gene ID: 3386. Target Synonyms: CD242; LW; Landsteiner-Weiner Blood System; Landsteiner-Wiener blood group glycoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8919