Learn More
Biomatik Corporation Human Interferon Alpha 10 (IFNa10) ELISA Kit
Supplier:  Biomatik Corporation EKU05081
Encompass_Preferred
Human Interferon Alpha 10 (IFNa10) ELISA Kit for detecting IFNa10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.62-1000pg/ml. Sensitivity: 26.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01566. Gene ID: 3446. Target Synonyms: LeIF C; Interferon alpha-C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7658