Learn More
Biomatik Corporation Human Interferon Alpha 10 (IFNa10) CLIA Kit
Supplier:  Biomatik Corporation EKU09647
Encompass_Preferred
Human Interferon Alpha 10 (IFNa10) CLIA Kit for detecting IFNa10 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.61pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P01566. Gene ID: 3446. Target Synonyms: LeIF C; Interferon alpha-C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7256