Learn More
Biomatik Corporation Human Interferon Alpha 2 (IFNa2) CLIA Kit
Supplier:  Biomatik Corporation EKU09644
Encompass_Preferred
Human Interferon Alpha 2 (IFNa2) CLIA Kit for detecting IFNa2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.49pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P01563. Gene ID: 3440. Target Synonyms: IFNA2A; IFNA2B; IFNA2C; LeIF A; Alpha-2a Interferon; Interferon Alpha 2b; Interferon Alpha A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6342