Learn More
Biomatik Corporation Human Interferon Alpha 4 (IFNa4) CLIA Kit
Supplier:  Biomatik Corporation EKU09643
Encompass_Preferred
Human Interferon Alpha 4 (IFNa4) CLIA Kit for detecting IFNa4 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.26pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P05014. Gene ID: 3441. Target Synonyms: INFA4; Interferon Alpha-4B; Interferon Alpha-76; Interferon Alpha-M1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6288