Learn More
Creative Biomart Interferon alfa-n3
Catalog No. 502084231
Encompass_Preferred
Purified, natural (n is for natural) human interferon alpha proteins (consists of 3 forms or polymorphisms including 2a, 2b and 2c). 166 residues, some are glycosylated (MW range from 16 kD to 27 kD).
Catalog No. 50-208-4231 Supplier Creative Biomart Supplier No. THP0180
Purified, natural (n is for natural) human interferon alpha proteins (consists of 3 forms or polymorphisms including 2a, 2b and 2c). 166 residues, some are glycosylated (MW range from 16 kD to 27 kD).