Learn More
Creative Biomart Interferon beta-1b
Supplier:  Creative Biomart THP0103
Encompass_Preferred
Human interferon beta (165 residues), cysteine 17 is substituted with serine. Produced in E. coli, no carbohydrates, MW=18.5kD
Catalog No. 50-208-4154