Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Human Interferon-gamma Recombinant Protein, E. coli, Lyophilized, 20 μg
SDP

Catalog No. 502050773
Encompass

This recombinant human interferon-gamma (hIFN-gamma) is expressed in E. coli and supplied as a lyophilized protein. IFN-gamma is a critical cytokine involved in the innate and adaptive immune responses, playing essential roles in the activation of macrophages and NK cells, as well as the enhancement of MHC class I and II expression.

  • Greater than 95% purity determined by SDS-PAGE
  • Endotoxin level is ≤ 1 EU per 1 μg
  • Bioactivity determined in a virus protection assay with an ED50 ≤ 0.75 ng/mL
  • Expressed in E. coli
  • Lyophilized format for stability
  • Molecular weight of 17 kDa

Catalog No. 50-205-0773 Supplier Cell Signaling Technology Supplier No. 80385S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This recombinant human interferon-gamma (hIFN-gamma) is expressed in E. coli and supplied as a lyophilized protein. IFN-gamma is a critical cytokine involved in the innate and adaptive immune responses, playing essential roles in the activation of macrophages and NK cells, as well as the enhancement of MHC class I and II expression.

  • Greater than 95% purity determined by SDS-PAGE
  • Endotoxin level is ≤ 1 EU per 1 μg
  • Bioactivity determined in a virus protection assay with an ED50 ≤ 0.75 ng/mL
  • Expressed in E. coli
  • Lyophilized format for stability
  • Molecular weight of 17 kDa

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.