Learn More
Biomatik Corporation Human Interferon Gamma (IFNg) CLIA Kit
Supplier:  Biomatik Corporation EKU09440
Encompass_Preferred
Human Interferon Gamma (IFNg) CLIA Kit for detecting IFNg in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.55pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P01579. Gene ID: 3458. Target Synonyms: IFN-G; IFG; IFI; INFr; IFN, Immune Interferon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8519