Learn More
Biomatik Corporation Human Interferon Induced Protein 44 (IFI44) ELISA Kit
Supplier:  Biomatik Corporation EKU11012
Encompass_Preferred
Human Interferon Induced Protein 44 (IFI44) ELISA Kit for detecting IFI44 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8TCB0. Gene ID: 10561. Target Synonyms: MTAP44; p44; Microtubule-associated protein 44. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4955