Learn More
American Research Products Inc Human interferon inducible pro
Catalog No. 5000326558
Encompass
Human interferon-inducible protein 10 IP-10 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26558 Supplier American Research Products Inc Supplier No. CSBE08181HIS
Human interferon-inducible protein 10 IP-10 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich