Learn More
Biomatik Corporation Human Interferon Inducible Protein 35 (IFI35) ELISA Kit
Supplier:  Biomatik Corporation EKU08447
Encompass_Preferred
Human Interferon Inducible Protein 35 (IFI35) ELISA Kit for detecting IFI35 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P80217. Gene ID: 3430. Target Synonyms: IFP35. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4424