Learn More
Biomatik Corporation Human Interferon Kappa (IFNk) ELISA Kit
Supplier:  Biomatik Corporation EKU12209
Encompass_Preferred
Human Interferon Kappa (IFNk) ELISA Kit for detecting IFNk in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 3.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9P0W0. Gene ID: 56832. Target Synonyms: IFN-K. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6518