Learn More
Biomatik Corporation Human Interleukin 1 Epsilon (IL1e) CLIA Kit
Supplier:  Biomatik Corporation EKU09568
Encompass_Preferred
Human Interleukin 1 Epsilon (IL1e) CLIA Kit for detecting IL1e in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.64pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9UHA7. Gene ID: 27179. Target Synonyms: IL1F6; IL36a; FIL1; FIL1 (EPSILON) ; FIL1E; IL-1F6; IL1 (EPSILON) ; Interleukin 36, Alpha; Interleukin 1 Family, Member 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7441