Learn More
Biomatik Corporation Human Interleukin 1 Family, Member 9 (IL1F9) CLIA Kit
Supplier:  Biomatik Corporation EKU09713
Encompass_Preferred
Human Interleukin 1 Family, Member 9 (IL1F9) CLIA Kit for detecting IL1F9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.52pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9NZH8. Gene ID: 56300. Target Synonyms: IL-1F9; IL36g; IL-1H1; IL-1RP2; IL1E; IL1H1; IL1RP2; Interleukin 36, Gamma; Interleukin-1 Homolog 1; Interleukin-1 epsilon; Interleukin 1-Related Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4352