Learn More
Biomatik Corporation Human Interleukin 1 Receptor Associated Kinase 3 (IRAK3) ELISA Kit
Supplier:  Biomatik Corporation EKU05162
Encompass_Preferred
Human Interleukin 1 Receptor Associated Kinase 3 (IRAK3) ELISA Kit for detecting IRAK3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y616. Gene ID: 11213. Target Synonyms: IRAK-M; IRAKM; IL-1 receptor-associated kinase M. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7759