Learn More
Biomatik Corporation Human Interleukin 1 Receptor Accessory Protein Like Protein 2 (IL1RAPL2) ELISA Kit
Supplier:  Biomatik Corporation EKU05152
Encompass_Preferred
Human Interleukin 1 Receptor Accessory Protein Like Protein 2 (IL1RAPL2) ELISA Kit for detecting IL1RAPL2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NP60. Gene ID: 26280. Target Synonyms: IL-1R9; IL1R9; IL1RAPL-2; TIGIRR-1; IL1RAPL-2-related protein; Interleukin-1 receptor 9; Three immunoglobulin domain-containing IL-1 receptor-related 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8917