Learn More
Biomatik Corporation Human Interleukin 1 Receptor Accessory Protein (IL1RAP) CLIA Kit
Supplier:  Biomatik Corporation EKU09649
Encompass_Preferred
Human Interleukin 1 Receptor Accessory Protein (IL1RAP) CLIA Kit for detecting IL1RAP in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 8.23-6,000pg/ml. Sensitivity: 3.91pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9NPH3. Gene ID: 3556. Target Synonyms: IL-1RAcP; C3orf13; IL1R3; Interleukin-1 receptor 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5765